Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Ficoll™400

Dissolves readily in aqueous solution, hydrophilic polymer of sucrose.

Manufacturer:  MilliporeSigma™ 455050GM

Catalog No. 45-505-0GM


Add to cart

Description

Description

EMD Millipore Calbiochem™ OmniPur™ Ficoll™400 is neutral highly branched.

  • Molecular biology grade
  • Specifications:
  • DNase: None Detected
  • Loss on drying: ≤5%
  • Protease: None Detected
  • RNase: None Detected
  • Color: White
Specifications

Specifications

Ficoll 400, Powder
50g
Molecular Biology Grade
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit