Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ Guanidine Isothiocyanate, Molecular Biology Grade

Inactivates RNases during isolation of RNA from cell lysate.

$228.50 - $802.00

Specifications

Product Type Guanidine Isothiocyanate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
51-401-KG
View Documents
MilliporeSigma™
51401KG
1kg
Each for $802.00
Add to Cart
 
51-202-50GM
View Documents
MilliporeSigma™
5120250GM
250g
Each for $228.50
Add to Cart
 
Description

Description

EMD Millipore Calbiochem™ OmniPur™ Guanidine Isothiocyanate, Molecular Biology Grade is protein denaturant.

  • Molecular biology grade
  • Specifications:
  • A (280nm, 70%, water): ≤0.8AU
  • Assay: ≥99.0%
  • DNase: None Detected
  • Melting point: 118 to 121°C
  • Protease: None Detected
  • RNase: None Detected
  • Solubility (70%, water): To pass test
Specifications

Specifications

Guanidine Isothiocyanate
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit