Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ OmniPur™ PBS 10X Liquid Concentrate, Molecular Biology Grade

Makes 10L of 1X PBS Buffer from each liter.

$155.00 - $617.00

Specifications

pH 7.3 to 7.5 at 25°C
Color Clear, Colorless
Product Type PBS 10X Liquid Concentrate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
65-042-0L View Documents MilliporeSigma™
650420L
20L Each for $617.00
Add to cart
 
65-054-L View Documents MilliporeSigma™
65054L
4L Each for $155.00
Add to cart
 
Description

Description

  • Molecular Biology Grade
  • DNase: None Detected
  • pH (10%, water) at 25°C: 7.3 to 7.5
  • RNase: None Detected
  • Sterility: Passes test
Specifications

Specifications

7.3 to 7.5 at 25°C
Clear, Colorless
PBS 10X Liquid Concentrate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit