Learn More
MilliporeSigma™ Calbiochem™ p21-Activated Kinase Inhibitor II, PF-3758309
p21-Activated Kinase Inhibitor II, PF-3758309, CAS 898044-15-0, is a cell-permeable, reversible, potent inhibitor of p21-activated kinases (IC50 = 190 and 99nM against PAK1 and PAK3, respectively).
Supplier: MilliporeSigma™ 5.00613.0001
Description
A cell-permeable pyrrolopyrazole that acts as a potent inhibitor against p21-activated kinases (Ki in nM/[ATP] inμM = 13.7/200, 18.7/40, 18.1/80. 17.1/72, respectively, in PAK1, PAK4, PAK5, PAK6 kinase assays; IC50 = 190 and 99nM against PAK1 and PAK3, respectively) by directly targeting the kinase ATP-binding site in a reversible manner ( K d = 4.5nM using rhPAK4300-591 kinase domain). Inhibits PAK4-dependent cellular signaling (IC50 = 24.2nM against TNFα-stimulated NF-κB activity in HEK293T) and proliferation (IC50 = 0.24 and 27 nM, respectively, against HCT116 and A549 colonies formation) in vitro and the growth of human tumors in nude mice in vivo (10 to 25mg/kg; b.i.d. p.o).Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
Powder | |
5 mg | |
99% by HPLC | |
C25H30N8OS | |
DMSO |
Protect from light | |
Yellowish-white | |
PAK | |
490.6g/mol |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.