Learn More
MilliporeSigma™ Calbiochem™ p38 MAP Kinase Inhibitor IX
The p38 MAP Kinase Inhibitor IX, also referenced under CAS 630106-85-3, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 5061612MG
Catalog No. 50-616-12MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
C22H17N7O2
DMSO
Off-white solid
2mg
411.4g/mol