Learn More
MilliporeSigma™ Calbiochem™ p38 MAP Kinase Inhibitor V
The p38 MAP Kinase Inhibitor V, also referenced under CAS 271576-77-3, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 5061561MG
Catalog No. 50-615-61MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
90% by HPLC
Protect from Light
p38 MAP Kinase
C19H20ClN5
DMSO or EtOH
Light yellow solid
1mg
271576-77-3
353.9g/mol