Learn More
MilliporeSigma™ Calbiochem™ p97 ATPase Activity Inhibitor, DBeQ
The p97 ATPase Activity Inhibitor, DBeQ controls the biological activity of p97 ATPase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Manufacturer:  MilliporeSigma™ 50619010MG
Catalog No. 50-619-010MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
C22H20N4
DMSO or EtOH
White solid
10mg
340.4g/mol