Learn More
MilliporeSigma™ Calbiochem™ PD 145065
A highly potent but non-selective endothelin receptor antagonist [IC50= 4nM for the ETA
Manufacturer:  MilliporeSigma™ 513020500UG
Catalog No. 51-302-0500UG
Add to cart
Description
Description
A highly potent but non-selective endothelin receptor antagonist [IC50 = 4nM for the ETA receptor (rabbit renal artery vascular smooth muscle cells); IC50 = 15nM for the ETB receptor (rat cerebellum)].
Specifications
Specifications
98% by HPLC
500μg
153049-49-1
992.1g/mol
White solid
Endothelin receptor antagonist
C52H63N7O10·2Na
Phosphate Buffer, pH 7.5