Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ PI 3-K Inhibitor XVIII
An N-myristoylated cell-permeable PI 3-K p110β-derived Gβγ-binding peptide
Supplier: MilliporeSigma™ 5.00499.0001
Description
An N-myristoylated cell-permeable PI 3-K p110β-derived Gβγ-binding peptide (aa 514-537 in p110β C2-helical linker region) that prevents Gβγ-, but not RTK-, mediated p100β activation both in cell-free assays (1μM) and in cultures (30μM in p110β/p85/Akt/Gβγ-transfected HEK 293E cells) without affecting basal p100β activity, p110β-Rab5 interaction, nor Gβγ-dependent activation of adenylyl cyclase or p101-p110γ dimer. The ATP-competitive p110β inhibitior TGX-221 (Cat. No. 528113) can be used in conjunction for studying Gβγ dependency of p110β-mediated cellular functions.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
Hygroscopic, Protect from light | |
Solid | |
Supplied as a trifluoroacetate salt. | |
Gβγ subunit of G-proteins | |
2629.1g/mol |
10 mg | |
White | |
95% by HPLC | |
C118H202N32O35 | |
H2O |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title