Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem PKC 20-28, Cell-Permeable, Myristoylated
Pseudosubstrate sequence from protein kinase Cα and β.
Manufacturer: MilliporeSigma 476480500UG
Description
- Peptide Sequence: N-Myr-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-NH2
- Cell Permeable
Specifications
Supplied as a trifluoroacetate salt. | |
Lyophilized solid | |
500μg | |
C60H106N18O11 | |
Tris-HCl, pH 7.5 or H2O |
95% by HPLC | |
Hygroscopic | |
TPA activation of MARCKS phosphorylation in fibroblast primary cultures | |
1255.6g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title