Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2

Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC50 = 58 nM, 210 nM, & 2.12 μM for Akt1, Akt2, and Akt3, respectively).

$251.00 - $294.00

Specifications

Content And Storage Protect from light
Form Solid
Color Yellow
Description 95% by HPLC
Target Proteins Akt1, Akt2, Akt3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-401-81MG
View Documents
MilliporeSigma™
1240181MG
1 mg
Each for $251.00
Add to cart
 
12-401-85MG
View Documents
MilliporeSigma™
1240185MG
5 mg
Each for $294.00
Add to cart
 
Description

Description

  • Packaged Under Inert Gas
  • Cell Permeable
Specifications

Specifications

Protect from light
Yellow
Akt1, Akt2, Akt3
C34H29N7O
DMSO
Solid
95% by HPLC
612847-09-3
551.6g/mol
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit