EMD Millipore Calbiochem™ PPACK, Dihydrochloride

PPACK, Dihydrochloride, CAS 142036-63-3, is an extremely potent and selective irreversible inhibitor of thrombin (Kobs/[I] = 107M-1S-1). Reacts with thrombin in a 1:1 stoichiometry.

Manufacturer: EMD Millipore 52022225MG

 View more versions of this product

Catalog No. 52-022-225MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Hygroscopic
    Protect from Light
    Protect from Moisture
    Target Proteins Thrombin
    Molecular Formula C21H33N6O3Cl3
    Formula Weight 523.9g/mol
    Solubility H2O
    Description 80% by HPLC
    Format White solid
    Quantity 25mg
    CAS 142036-63-3

    • Packaged Under Inert Gas
    • Peptide Sequence: H-D-Phe-Pro-Arg-chloromethylketone, 2HCl

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit