Learn More
EMD Millipore Calbiochem™ Protease Inhibitor Cocktail Set VII Available on GSA/VA Contract for Federal Government customers only.
Protease Inhibitor Cocktail Set VII contains five protease inhibitors with broad specificity. Recommended for purification of proteins containing His·Tag® sequences.
Manufacturer:  EMD Millipore 5391381ML
 View more versions of this product
Catalog No. 53-913-81ML
  Check Availability
Add to cart
Specifications
Description
A cocktail of five protease inhibitors with broad specificity for the inhibition of cysteine, serine, aspartic, and thermolysin-like proteases and aminopeptidases. This cocktail is recommended for purification of proteins containing His Tag® sequences. Each vial contains 100mm AEBSF, HCl (Cat. No. 101500), 5mm Bestatin (Cat. No. 200484), 1.5mm E-64 (Cat. No. 324890), 2mm Pepstatin A (Cat. No. 516481), and 200µM of Phosphoramidon, Disodium Salt (Cat. No. 525276). OnemL is recommended for the inhibition of proteases in 10 g of cells. Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
Description & Specifications
Specifications
In 1mL DMSO.
Liquid
1mL