Learn More
MilliporeSigma™ Calbiochem™ (R)-CPPene Available on GSA/VA Contract for Federal Government customers only.
A highly potent competitive antagonist of NMDA receptors (ED50 = 39nM; pA2
Manufacturer:  MilliporeSigma™ 5.04840.0001
Catalog No. 50-484-00001
Add to cart
Description
Description
A highly potent competitive antagonist of NMDA receptors (ED50 = 39nM; pA2 = 6.8). Biologically active admitted systemically. Frequently used in assessing the functional roles of NMDA receptors mediated neurotransmissions in a variety of research areas, such as excitotoxic cell death, apoptosis, bipolar disorders, ischemia, traumatic brain injury, epilepsy, and neuropathic pain.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
99% by HPLC
Protect from Light
NMDA receptors
H2O
White solid
10mg
C8H15N2O5P