Learn More
MilliporeSigma™ Calbiochem™ RapidStep™ ECL Reagent
Detects immobilized proteins, labeled with horseradish peroxidase by immunoblotting.
Supplier:  MilliporeSigma™ 345818100ML
Catalog No. 34-581-8100ML
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
EMD Millipore Calbiochem™ RapidStep™ ECL Reagent is single component reagent.
  • Chemiluminescent detection
  • Contains luminol enhancer combined in one solutionReady to use, convenient, cost-effective
  • Packaging: Glass Bottle
Specifications
Specifications
ECL Western Blot Detection Reagent
100 mL
2°C to 8°C
Recommendations
Recommendations