Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Ras-Net (Elk-3) Pathway Inhibitor, XRP44X
The Ras-Net (Elk-3) Pathway Inhibitor, XRP44X, also referenced under CAS 729605-21-4, controls the biological activity of Ras-Net (Elk-3).
Supplier: MilliporeSigma™ 4801505MG
Description
Packaged Under Inert GasSpecifications
White | |
729605-21-4 | |
380.9g/mol | |
Protect from light | |
5 mg |
98% by HPLC | |
C21H21ClN4O | |
DMSO or EtOH | |
Solid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction