Learn More
MilliporeSigma™ Calbiochem™ Ro 25-6981 Maleate Salt
A potent, activity-dependent antagonist highly selective for GluN1/NR2B subtype NMDA receptors (IC USB>50 =
Manufacturer:  MilliporeSigma™ 5.04845.0001
Catalog No. 50-484-50001
Add to cart
Description
Description
A potent, activity-dependent antagonist highly selective for GluN1/NR2B subtype NMDA receptors (IC USB>50 = 0.009 and 52µM for NR2B and NR2A subtype receptors respectively). Commonly used for studying brain functions and disorders involving neurotransmissions mediated by GluN1/NR2B subtype NMDA receptors (learning and memory, neuroplasticity, neurotoxicity, neuropathic pain, addiction, and neurogenesis enhancement).
Specifications
Specifications
99% by Chiral HPLC
Protect from Light
GluN1/NR2B receptors
H2O
White solid
1mg
C22H29NO2·C4H4O4