EMD Millipore Calbiochem™ Roscovitine

A potent, reversible, and selective inhibitor of Cdks that exhibits about 10-fold greater efficacy towards p34-cdk1 and p33-cdk2 and 20-fold greater efficacy towards p33-cdk5 relative to Olomoucine.

Manufacturer: EMD Millipore 5573601MG

 View more versions of this product

Catalog No. 55-736-01MG

  Check Availability
Add to Cart

    Specifications

    Target Proteins p34cdk1/cyclin B
    Molecular Formula C19H26N6O
    Formula Weight 354.5g/mol
    Solubility DMSO
    Description 95% by HPLC
    Format White to off-white solid
    Quantity 1mg
    CAS 186692-46-6

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit