Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ RXRα Inhibitor, K-80003
Click to view available options
Quantity:
10 mg
Description
A cell permeable, Sulindac analog with IC50=2.4μM for RXRα binding, and unlike other NSAIDs, is inactive toward COX2 and COX1 even at 1 mM. The compound displays potent inhibition of RXRα dependent AKT Activation in both PC3 and A549 cells, and is RXRα but not RXRγ expression dependent as shown in PC3 cells. It is further shown to inhibit the interaction of RXRα/δ80 with p85α either in the absence or presence of TNF-α. Induces PARP cleavage and caspase-8 activation at 50μM when used with TNF-α. Furthermore, it almost completely suppresses colony formation of HeLa/RXRα/1-134 and RXRα/δ80 cells at 25μM in vitro, and exhibits potent inhibition of tumor growth in vivo when dosed at 60mg/kg.

Specifications
Specifications
Color | Yellow |
Description | 95% by HPLC |
Molecular Formula | C23H23FO2 |
Formula Weight | 350.4g/mol |
Solubility | DMSO |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction