Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ (S)-5-Iodowillardiine

A potent and selective agonist for GluK1-containing kainate receptors (EC50 = 0.06 and 0.21 μM for GluK1/K5 and GluK1, respectively). Does not interact with GluK2, GluK6, and GluK7 receptors.

Manufacturer:  MilliporeSigma™ 5.09707.0001

Catalog No. 50-970-70001

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
Specifications

Specifications

10 mg
White
GluK1
C7H8IN3O4
Solid
98% by HPLC
140187-25-3
1N NaOH
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit