Learn More
MilliporeSigma™ Calbiochem™ S18986
A positive allosteric modulator selective for the AMPA receptors
Manufacturer:  MilliporeSigma™ 5.04983.0001
Catalog No. 50-498-30001
Add to cart
Description
Description
A positive allosteric modulator selective for the AMPA receptors. A nootropic and neuroprotective agents frequently used in researches studying cognition, memory, and aging related cognitive declines. Induces production of BNDF and AMPA-mediated release of noradrenaline and acetylcholine. Biologically viable admitted systematically.
Specifications
Specifications
97% by HPLC
Protect from Light
AMPA
224.28g/mol
White solid
5mg
C10H12N2O2S
DMSO