Learn More
MilliporeSigma™ Calbiochem™ Salmeterol Xinafoate
A potent and selective β2 -adrenoceptor agonist (EC50
Manufacturer:  MilliporeSigma™ 5.04815.0001
Catalog No. 50-481-50001
Add to cart
Description
Description
A potent and selective β2-adrenoceptor agonist (EC50 = 5.3nM) that displays bronchodilator actions. Unlike other β2- adrenoceptor agonists, Salmeterol binds to exo-site domain of β2-AR receptors, producing a slow onset of action and prolonged activation. Clinically used for the treatment of asthma and chronic obstructive pulmonary disease(COPD).
Specifications
Specifications
98% by HPLC
Protect from Light
→2-adrenoceptor
603.75g/mol
White powder
10mg
C25H37NO4·C11H8O3
MeOH, EtOH, or DMSO