EMD Millipore Calbiochem™ SB 328437

A highly potent and selective CC chemokine receptor-3 (CCR3) antagonist.

Manufacturer: EMD Millipore 5594061MG

 View more versions of this product

Catalog No. 55-940-61MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins CC chemokine receptor-3 (CCR3)
    Molecular Formula C21H18N2O5
    Formula Weight 378.4g/mol
    Solubility DMSO
    Description 98% by HPLC
    Format White solid
    Quantity 1mg
    CAS 247580-43-4

    Packaged Under Inert Gas

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit