EMD Millipore Calbiochem™ γ-Secretase Inhibitor XX

This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by »72% in Tg2576 mutant APP transgenic mouse model.

Manufacturer: EMD Millipore 5657891MG

 View more versions of this product

Catalog No. 56-578-91MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Light
    Target Proteins γ-secretase
    Molecular Formula C26H23F2N3O3
    Formula Weight 463.4g/mol
    Solubility DMSO
    Description 95% by HPLC
    Format White solid
    Quantity 1mg
    CAS 209984-56-5

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit