Learn More
MilliporeSigma™ Calbiochem™ Sialyl Lewis x, Sodium Salt
Acts as minimal binding lig for E-selectins.
$355.00 - $1161.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
56-595-01MG
|
MilliporeSigma™
5659501MG |
1 mg |
Each for $355.00
|
|
|||||
56-595-05MG
|
MilliporeSigma™
5659505MG |
5 mg |
Each for $1,161.00
|
|
|||||
Description
EMD Millipore Calbiochem™ Sialyl Lewis x, Sodium Salt is an enzymatically prepared tetrasaccharide.
- Reduces injury, ischemia, reperfusion, immune complex-induced acute lung injury in animal models
- Blocks E-selectin-mediated neutrophil recruitment to sites of inflammation in animal models
Specifications
-20°C |