Learn More
MilliporeSigma™ Calbiochem™ Sphingosine Kinase Inhibitor
The Sphingosine Kinase Inhibitor, also referenced under CAS 1177741-83-1, controls the biological activity of Sphingosine Kinase. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Supplier: MilliporeSigma™ 5.00489.0001
Description
Packaged Under Inert GasSpecifications
Protect from light | |
Powder | |
98% by HPLC | |
C18H18ClNO3 | |
DMSO |
10 mg | |
Yellow | |
1177741-83-1 | |
331.8g/mol |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.