EMD Millipore Calbiochem™ STAT3 Inhibitor Peptide, Cell-Permeable

The STAT3 Inhibitor Peptide, Cell-Permeable controls the biological activity of STAT3.

Manufacturer: EMD Millipore 5730961MG

 View more versions of this product

Catalog No. 57-309-61MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Hygroscopic
    Target Proteins Blocker of Stat3 activation
    Molecular Formula C92H157N20O24P
    Formula Weight 1958.4g/mol
    Solubility H2O
    Formulation Supplied as a trifluoroacetate salt.
    Description 95% by HPLC
    Format White lyophilized solid
    Quantity 1mg

    • Packaged Under Inert Gas
    • Peptide Sequence: H-Pro-Tyr-(PO3H2)-Leu-Lys-Thr-Lys-Ala-Ala-Val-Leu-Leu-Pro-Val-Leu-Leu-Ala-Ala-Pro-OH
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit