Learn More
MilliporeSigma™ Calbiochem™ Adenylyl Cyclase Type V Inhibitor, NKY80
The Adenylyl Cyclase Type V Inhibitor, NKY80, also referenced under CAS 299442-43-6, controls the biological activity of Adenylyl Cyclase Type V.
Manufacturer:  MilliporeSigma™ 11685025MG
 View more versions of this product
Catalog No. 11-685-025MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
97% by HPLC
Protect from Light
Adenylate cyclase (AC) type V isoform
C12H11N3O2
DMSO or MeOH
Pale orange solid
25mg
299442-43-6
229.2g/mol