Learn More
EMD Millipore Calbiochem™ Suramin, Sodium Salt
A reversible and competitive inhibitor of protein tyrosine phosphatases.
Manufacturer:  EMD Millipore 574625200MG
 View more versions of this product
Catalog No. 57-462-5200MG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
98% by HPLC
Hygroscopic
protein tyrosine phosphatases
C51H34N6O23S6·6Na
H2O
White crystalline solid
200mg
129-46-4
1429.2g/mol