Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Survivin Inhibitor III, LLP-3
A cell-permeable, dihydropyridinone compound that selectively disrupts survivin-Ran interaction in cell-free binding assays (IC50 &l
Supplier: MilliporeSigma™ 5.00614.0001
Description
A cell-permeable, dihydropyridinone compound that selectively disrupts survivin-Ran interaction in cell-free binding assays (IC50 <0.3μM) and in U87 glioma cells (by 55% in 24 h; 20μM) via direct binding at survivin protein-protein interaction interface, while exhibiting much reduced or little potency against survivin-Smac/DIABLO, survivin-survivin, or XIAP-Smac/DIABLO interaction. LLP-3 treatment abrogates neurosphere formation in GMB cultures (IC50 ≤35μM) by selectively depleting CD133+ GSC population via apoptosis induction. TMZ (Cat. No. 500609), in comparison, enriches GMB GSC population by preferentially eradicating non-GSC population. LLP-3 is also efficacious in treating human GMB xenograft in mice in vivo (25mg/kg/day via i.p.).Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
10 mg | |
Protect from light | |
99% by HPLC | |
C32H23ClN2O4 | |
DMSO |
Solid | |
Yellow | |
survivin | |
535g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title