Learn More
MilliporeSigma™ Calbiochem™ Tamsulosin Hydrochloride
A selective α1A adrenoceptor antagonist that has preferential selectivity for the α1A adrenoceptor in the prostate versus the α1B adrenoceptor in the blood vessels (pKi
Manufacturer:  MilliporeSigma™ 5.04793.0001
Catalog No. 50-479-30001
Add to cart
Description
Description
A selective α1A adrenoceptor antagonist that has preferential selectivity for the α1A adrenoceptor in the prostate versus the α1B adrenoceptor in the blood vessels (pKi values are 9.97, 9.64 and 8.86 for α1A, α1B and α1D subtypes respectively). Used for treatment of difficult urination, a common symptom of enlarged prostate. Has negligible effect on arterial blood pressure in vivo .
Specifications
Specifications
98% by HPLC
Hygroscopic
Protect from Light
Protect from Moisture
→1A adrenoceptor
444.97g/mol
White powder
10mg
C20H28N2O5S·HCl
DMSO