Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Thyroid Hormone Receptor Antagonist, 1-850
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | Yellow |
| Description | 95% by HPLC |
| Target Proteins | Thyroid Hormone Receptor |
| CAS | 251310-57-3 |
| Molecular Formula | C21H20F3N5O4 |
| Formula Weight | 463.4g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction