Learn More
MilliporeSigma™ Calbiochem™ Z-Guggulsterone
A synthetic form of the guggul tree plant steroid that exhibits an ability to lower LDL cholesterol and triglyceride levels.
Supplier: MilliporeSigma™ 37069025MG
Description
Packaged Under Inert GasSpecifications
White | |
Farnesoid X receptor (FXR) | |
312.5g/mol | |
Solid |
Single spot by TLC | |
C21H28O2 | |
DMSO | |
25 mg |