EMD Millipore Calbiochem™ Z-Guggulsterone

A synthetic form of the guggul tree plant steroid that exhibits an ability to lower LDL cholesterol and triglyceride levels.

Manufacturer: EMD Millipore 37069010MG

 View more versions of this product

Catalog No. 37-069-010MG

  Check Availability
Add to Cart

    Specifications

    Target Proteins Farnesoid X receptor (FXR)
    Molecular Formula C21H28O2
    Formula Weight 312.5g/mol
    Solubility DMSO
    Description Single spot by TLC
    Format White solid
    Quantity 10mg

    Packaged Under Inert Gas

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit