Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Calcineurin A Mouse anti-Human, Clone: BF1.1, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Calcineurin A
Clone BF1.1
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122788
View Documents
Novus Biologicals
NBP30765720UG
20 μg
Each for $138.37
N/A
NB122787
View Documents
Novus Biologicals
NBP30765710UG
100 μg
Each for $352.89
N/A
Description

Description

Calcineurin A Monoclonal antibody specifically detects Calcineurin A in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

Calcineurin A
0.2 mg/ml
Western Blot, ELISA
Purified
RUO
Human
calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Recombinant full-length Calcineurin A protein
Monoclonal
Protein A or G purified
BF1.1
Western Blot 1-2 ug/ml, ELISA
Unconjugated
Mouse
Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
5530
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit