Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcion, Acros Organics

Click to view available options
Quantity:
1g
3810-39-7
C30H14N4Na6O22S6
1112.747
RZTLWYLHENJEJY-ZUZCPLCPSA-H
sodium 5-1,8-dihydroxy-3,6-disulphonaphthalen-2-yl azo-4-hydroxy-3 8-hydroxy-3,6-disulphonaphthalen-1-yl azo naphthalene-2,7-disulphonate,5-1,8-dihydroxy-3,6-disulfo-2-naphthyl-azo-4-hydroxy-3-8-hydroxy-3,6-disulfo-1-naphthyl-azo-2,7-naphthalenedisul,2,7-naphthalenedisulfonic acid, 5-1,8-dihydroxy-3,6-disulfo-2-naphthalenyl azo-4-hydroxy-3-8-hydroxy-3,6-disulfo-1-naphthalenyl azo-, hexasodium salt,2,7-naphthalenedisulfonic acid, 5-2-1,8-dihydroxy-3,6-disulfo-2-naphthalenyl diazenyl-4-hydroxy-3-2-8-hydroxy-3,6-disulfo-1-naphthalenyl diazenyl-, sodium salt 1:6,hexapotassium 4-hydroxy-5-2-8-2-8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene hydrazin-1-yl-1-oxo-3,6-disulfonatonaphthalen-2-ylidene hydrazin-1-yl naphthalene-2,7-disulfonate
9589349
hexasodium;(3Z)-3-[(8-hydroxy-3,6-disulfonatonaphthalen-1-yl)hydrazinylidene]-5-[(2Z)-2-(8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-4-oxonaphthalene-2,7-disulfonate
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)[O-])NN=C3C(=CC4=CC(=CC(=C4C3=O)NN=C5C(=CC6=CC(=CC(=C6C5=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
3810-39-7
C30H14N4Na6O22S6
1112.747
RZTLWYLHENJEJY-ZUZCPLCPSA-H
sodium 5-1,8-dihydroxy-3,6-disulphonaphthalen-2-yl azo-4-hydroxy-3 8-hydroxy-3,6-disulphonaphthalen-1-yl azo naphthalene-2,7-disulphonate,5-1,8-dihydroxy-3,6-disulfo-2-naphthyl-azo-4-hydroxy-3-8-hydroxy-3,6-disulfo-1-naphthyl-azo-2,7-naphthalenedisul,2,7-naphthalenedisulfonic acid, 5-1,8-dihydroxy-3,6-disulfo-2-naphthalenyl azo-4-hydroxy-3-8-hydroxy-3,6-disulfo-1-naphthalenyl azo-, hexasodium salt,2,7-naphthalenedisulfonic acid, 5-2-1,8-dihydroxy-3,6-disulfo-2-naphthalenyl diazenyl-4-hydroxy-3-2-8-hydroxy-3,6-disulfo-1-naphthalenyl diazenyl-, sodium salt 1:6,hexapotassium 4-hydroxy-5-2-8-2-8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene hydrazin-1-yl-1-oxo-3,6-disulfonatonaphthalen-2-ylidene hydrazin-1-yl naphthalene-2,7-disulfonate
9589349
hexasodium;(3Z)-3-[(8-hydroxy-3,6-disulfonatonaphthalen-1-yl)hydrazinylidene]-5-[(2Z)-2-(8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene)hydrazinyl]-4-oxonaphthalene-2,7-disulfonate
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)[O-])NN=C3C(=CC4=CC(=CC(=C4C3=O)NN=C5C(=CC6=CC(=CC(=C6C5=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
Quantity 1g
Formula Weight 1112.78
Chemical Name or Material Calcion