Calcite, White pieces, Alfa Aesar™

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Manufacturer: Alfa Aesar 44520-04

Quantity 2g

 View more versions of this product

Catalog No. AA4452004

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 


    Chemical Identifiers

    CAS 13397-26-7
    Physical Form White pieces
    Molecular Formula CCaO3
    Formula Weight 100.09g/mol
    MDL Number MFCD00010906
    Synonym Calcium Carbonate (naturally occuring mineral)
    Chemical Name or Material Calcite

    Specifications

    Density 2.93g/cm3 at 20°C
    Color White
    Quantity 2g
    CAS 13397-26-7
    Physical Form White pieces
    Molecular Formula CCaO3
    Formula Weight 100.09g/mol
    MDL Number MFCD00010906
    Synonym Calcium Carbonate (naturally occuring mineral)
    Chemical Name or Material Calcite

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit