Learn More
Alfa Aesar™ Calcitonin (8-32), Salmon
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J64415EXD
 View more versions of this product
Catalog No. AAJ64415EXD
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
2725.06
Salmon
Solid
RUO
Calcitonin (8-32), salmon
HPLC
0.5mg
Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2
-30° to -10°C