Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Calcitonin Ab-2, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB1519P0

 View more versions of this product

Catalog No. RB1519P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Calcitonin Ab-2, Rabbit Polyclonal Antibody.

Calcitonin is demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.

Immunogen: A synthetic peptide from human calcitonin

Positive Control: Thyroid or medullary CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Calcitonin Ab-2
Polyclonal
1mg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
RUO
Primary
Polyclonal
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
Concentrated
A synthetic peptide from human calcitonin
100μL
Steroids Hormones and Growth Factors
Primary
Canine, Equine, Human, Murine, Non-human Primate, Porcine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit