Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Calcium 2-ethylhexanoate, Superconductor Grade, in 2-ethylhexanoic acid (3-8% Ca)
Click to view available options
Quantity:
5g
25g
100g
Specifications
Concentration or Composition (by Analyte or Components) | 3 to 8% Ca |
Odor | Weak, characteristic |
Quantity | 100g |
Formula Weight | 326.5 |
Physical Form | Liquid |
Grade | Superconductor |
Chemical Name or Material | Calcium 2-ethylhexanoate, in 2-ethylhexanoic acid |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H361d
EINECSNumber : 205-249-0
TSCA : Yes
Recommended Storage : Ambient temperatures