Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Acetate, EP, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
62-54-4
C4H6CaO4
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
[Ca++].CC([O-])=O.CC([O-])=O
62-54-4
C4H6CaO4
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
[Ca++].CC([O-])=O.CC([O-])=O

C1067, 62-54-4, C4H6CaO4

Spectrum™ Calcium Acetate, USP, EP is used to prevent high blood phosphate levels in patients who are on dialysis due to severe kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Arsenic (As) 3 ppm
Percent Purity 99-100.5%
Chloride 330 ppm
Fluoride 50 ppm
Lead (Pb) 0.001%
Packaging Poly Pail
pH Range 7.2-8.2
Sulfate 600 ppm
Water 7%
Quantity 12 kg
Show More Show Less