Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Acetate, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

Chemical Identifiers

5743-26-0
C4H8CaO5
176.181
XQKKWWCELHKGKB-UHFFFAOYSA-L
82163
CHEBI:59199
calcium;diacetate;hydrate
CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 176.19
Physical Form Powder
Packaging Poly Pail
Specific Gravity 1L = 1.50kg
Quantity 2.5kg
Chemical Name or Material Calcium Acetate, Monohydrate
Grade Baker Analyzed™ ACS Reagent