Learn More
Calcium Acetate, Powder, FCC, 99-100.5%, Spectrum™
Click to view available options
Packaging:
Fiber Drum
Poly Pail
CA128, 62-54-4, C4H6CaO4
Spectrum™ Calcium Acetate, Powder, FCC is used as a stabilizer in most hard candies and as an anti-roping agent in most commercially sold breads. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chloride 0.0005
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Water 0.07