Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Acetate, Powder, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Pail
62-54-4
C4H6CaO4
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
[Ca++].CC([O-])=O.CC([O-])=O
62-54-4
C4H6CaO4
158.17
VSGNNIFQASZAOI-UHFFFAOYSA-L
calcium diacetate
[Ca++].CC([O-])=O.CC([O-])=O

CA128, 62-54-4, C4H6CaO4

Spectrum™ Calcium Acetate, Powder, FCC is used as a stabilizer in most hard candies and as an anti-roping agent in most commercially sold breads. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Chloride 0.05%
Percent Purity 99-100.5%
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Water 7%
Quantity 2.5 kg