Alfa Aesar™ Calcium alpha-D-isosaccharinate, 98% (Assay)

Manufacturer: Alfa Aesar™ 4437106

Quantity 5g

 View more versions of this product

Catalog No. AA4437106

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C12H22CaO12
    Linear Formula [CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
    Formula Weight 398.38
    MDL Number MFCD04039767
    Chemical Name or Material Calcium alpha-D-isosaccharinate

    Specifications

    Formula Weight 398.38
    Physical Form Crystalline
    Molecular Formula C12H22CaO12
    MDL Number MFCD04039767
    Quantity 5g
    Linear Formula [CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
    Color Pale brown
    Chemical Name or Material Calcium alpha-D-isosaccharinate
    Assay 98%, (Assay)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit