Alfa Aesar™ Calcium bis(trifluoromethylsulfonyl)imide

Manufacturer: Alfa Aesar™ 46533MD

Quantity 250mg

 View more versions of this product

Catalog No. AA46533MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 165324-09-4
    Molecular Formula C4CaF12N2O8S4
    Linear Formula [(CF3SO2)2N]2Ca
    Formula Weight 600.35
    MDL Number MFCD14156019
    UN Number UN1759
    Synonym Calcium triflimide; Calcium bis(trifluoromethanesulfonimide)
    Chemical Name or Material Calcium bis (trifluoromethylsulfonyl)imide

    Specifications

    Formula Weight 600.35
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    MDL Number MFCD14156019
    Linear Formula [(CF3SO2)2N]2Ca
    UN Number UN1759
    Synonym Calcium triflimide; Calcium bis(trifluoromethanesulfonimide)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 250mg
    Molecular Formula C4CaF12N2O8S4
    CAS 165324-09-4
    Melting Point >300°C
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Chemical Name or Material Calcium bis (trifluoromethylsulfonyl)imide

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit