Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium bromide hydrate, extra pure, Acros Organics

Click to view available options
Quantity:
250g
2.5kg
71626-99-8
Br2Ca·xH2O
199.9
71626-99-8
Br2Ca·xH2O
199.9
Linear Formula CaBr2·xH2O
Solubility Information Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol,ethanol
Formula Weight 199.9
Grade Extra Pure
Packaging Plastic bottle
Solubility (10% in water) Clear colorless to slightly hazy
Melting Point 730.0°C
Quantity 2.5kg
Chemical Name or Material Calcium bromide hydrate

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger