Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbide, 72-82%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Glass bottle
75-20-7
C2Ca
64.1
MFCD00010905
calcium carbide,3-phenoxypropylene di acetate,glycerol phenyl ether diacetate,3-phenoxy-1,2-propanediol diacetate,phenylglyceryl ether diacetate,1-phenyl ether-2,3-diacetate glycerol,1,2-propanediol, 3-phenoxy-, diacetate,1-acetyloxy-3-phenoxypropan-2-yl acetate,quartz crystal,3-phenoxypropane-1,2-diyl diacetate
75-20-7
C2Ca
64.1
MFCD00010905
calcium carbide,3-phenoxypropylene di acetate,glycerol phenyl ether diacetate,3-phenoxy-1,2-propanediol diacetate,phenylglyceryl ether diacetate,1-phenyl ether-2,3-diacetate glycerol,1,2-propanediol, 3-phenoxy-, diacetate,1-acetyloxy-3-phenoxypropan-2-yl acetate,quartz crystal,3-phenoxypropane-1,2-diyl diacetate
Assay Percent Range 72-82%
Linear Formula CaC2
Merck Index 15, 1658
Solubility Information Solubility in water: hydrolyses
Formula Weight 64.1
Physical Form Lumps
Percent Purity 72-82%
Identification Positive
Packaging Glass bottle
Solubility Solubility in water: hydrolyses.
Boiling Point 2300.0°C
Color Gray
Melting Point 2160.0°C
Quantity 250g
Chemical Name or Material Calcium carbide
Show More Show Less

GHS H Statement:
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger