Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbide, Alfa Aesar™

Manufacturer:  Alfa Aesar 14308A1

Quantity 1kg

 View more versions of this product

Catalog No. AA14308A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

75-20-7
C2Ca
MFCD00010905
Calcium carbide
9 to 40mm Lump
64.09
Calcium acetylide

Specifications

75-20-7
C2Ca
64.09
Calcium acetylide
Gray-black
Calcium carbide
9 to 40mm Lump
Grayish black lump
MFCD00010905
2.22g/cm3
1kg

Safety and Handling

SDS & Safety Documents

H261-H314

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 200-848-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit