Calcium carbonate, 99.997% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 4307314

Quantity 25g

 View more versions of this product

Catalog No. AA4307314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 471-34-1
    Physical Form Powder
    Assay Percent Range 99.997% (Metal basis)
    Molecular Formula CaCO3
    Formula Weight 100.09
    MDL Number MFCD00010906
    Chemical Name or Material Calcium carbonate

    Specifications

    Formula Weight 100.09
    Quantity 25g
    Color White
    Physical Form Powder
    Assay Percent Range 99.997% (Metal basis)
    Molecular Formula CaCO3
    Appearance White powder
    MDL Number MFCD00010906
    CAS 471-34-1
    Chemical Name or Material Calcium carbonate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit